
Benzyloxyacetaldehyde diethyl acetal 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:42783-78-8
:
Formula:C₁₃H₂₀O₃
MDL Number:MFCD00051786
Boiling Pt:99…100 °C
Density:0.987 g/cm³ (25 °C)
Flash Pt:79 °C
:224.30 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together