4-Spiroindanepiperidine hydrochloride
Catalog # APOSOR451148-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:223.75 g/mol
Storage Temperature:Ambient
CAS Number:96651-85-3
Formula:C₁₃H₁₈ClN
MDL Number:MFCD08461061