tert-Butyl-3-(1-hydroxy-1-methylethyl)azetidine-1-carboxylate
Catalog # APOSOR451036-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:215.29 g/mol
Storage Temperature:Ambient
CAS Number:1257293-79-0
Formula:C₁₁H₂₁NO₃
MDL Number:MFCD22370133