
Pyridine-3-sulphonyl chloride hydrochloride, Technical Grade
Catalog # APOSOR4393-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:144…145 °C
Storage Temperature:Freezer
UN:3261
ADR:8,II
:GHS05
CAS Number:42899-76-3
Formula:C5H5Cl2NO2S
MDL Number:MFCD04035552