3,5-Dimethylbenzoyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6613-44-1
:
Formula:C₉H₉ClO
MDL Number:MFCD02313742
:168.62 g/mol
Boiling Pt:127 °C/20mmHg
Density:1.14 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:413-010-9
UN:3265
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together