
6-Chloropyrazine-2-carboxylic acid
Catalog # APOSOR40053-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:158.54 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:23688-89-3
Formula:C₅H₃ClN₂O₂
MDL Number:MFCD00837871