Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-6-chloroanisole
2-Bromo-6-chloroanisole
2-Bromo-6-chloroanisole
APOSOR400467-1G
:
Apollo Scientific
:
2-Bromo-6-chloroanisole
APOSOR400467-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
221.48 g/mol
:
GHS07
:
Ambient
:
174913-10-1
:
C₇H₆BrClO
:
MFCD15143586