Methyl-3-amino-4-methylbenzoate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18595-18-1
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00025206
:165.19 g/mol
Melting Pt:113…117 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together