2-Methyl-2H-indazol-6-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:50593-30-1
:
Formula:C₈H₉N₃
MDL Number:MFCD00464866
:147.18 g/mol
Melting Pt:148…150 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together