Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-5-formyl-3-methylisoxazole-4-carboxylate
Ethyl-5-formyl-3-methylisoxazole-4-carboxylate
Ethyl-5-formyl-3-methylisoxazole-4-carboxylate
APOSOR346092-1G
:
Apollo Scientific
:
Ethyl-5-formyl-3-methylisoxazole-4-carboxylate
APOSOR346092-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
183.17 g/mol
:
GHS07
:
Ambient
:
129663-12-3
:
C₈H₉NO₄
:
MFCD10568350