Order Entry
United Kingdom
ContactUsLinkComponent
3-Methoxy-5-methylbenzoic acid 97%
3-Methoxy-5-methylbenzoic acid 97%
Catalog # APOSOR345381-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Methoxy-5-methylbenzoic acid 97%
Catalog # APOSOR345381-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :166.1700 g/mol
Storage Temperature:Ambient
CAS Number:62089-34-3
Formula:C9H10O3
MDL Number:MFCD12405314