6-Methoxy-2H-chromene-3-carbaldehyde 95%
Catalog # APOSOR33415-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:190.2 g/mol
Storage Temperature:Ambient
CAS Number:57543-40-5
Formula:C₁₁H₁₀O₃
MDL Number:MFCD01662418