
2,4,6-Trichlorophenylboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:73852-18-3
:
Formula:C₆H₄BCl₃O₂
MDL Number:MFCD01074621
:225.26 g/mol
Melting Pt:162…164 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together