(2-Chloro-1,3-thiazol-5-yl)methyl phenyl ether 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:339018-23-4
:
Formula:C₁₀H₈NOClS
MDL Number:MFCD01443853
:225.6946 g/mol
Storage Temperature:Ambient
Frequently Bought Together