5-(4-Chlorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:35982-98-0
:
Formula:C₁₂H₇ClN₂O
MDL Number:MFCD00231524
:230.66 g/mol
Storage Temperature:Ambient
Frequently Bought Together