
2-Bromo-5H-pyrrolo[2,3-b]pyrazine 97%
Catalog # APOSOR323262-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:198.0201 g/mol
Boiling Pt:265 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:875781-43-4
Formula:C₆H₄BrN₃
MDL Number:MFCD09834822