Order Entry
United Kingdom
ContactUsLinkComponent
Pyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione 95%
Pyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione 95%
Catalog # APOSOR322698-1G
Supplier:  Apollo Scientific
CAS Number:  
Pyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione 95%
Catalog # APOSOR322698-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :163.1335 g/mol
Storage Temperature:Ambient
CAS Number:21038-67-5
Formula:C7H5N3O2
MDL Number:MFCD07438024