Order Entry
United Kingdom
ContactUsLinkComponent
3-Chloropyrazine-2-carboxylic acid 98%
Irritant
3-Chloropyrazine-2-carboxylic acid 98%
Catalog # APOSOR322467-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Chloropyrazine-2-carboxylic acid 98%
Catalog # APOSOR322467-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:117…118 °C
 :158.5425 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:27398-39-6
Formula:C5H3ClN2O2
MDL Number:MFCD01318438