
Methyl-DL-2-aminobutyrate hydrochloride
Catalog # APOSOR322366-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:153.61 g/mol
Melting Pt:150 °C
:GHS07
Storage Temperature:Ambient
CAS Number:7682-18-0
Formula:C₅H₁₁NO₂
MDL Number:MFCD00058295