

3-Chlorophenyl isocyanate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2909-38-8
:
Formula:C₇H₄ClNO
MDL Number:MFCD00002016
:153.57 g/mol
Boiling Pt:201 °C/760mmHg
Melting Pt:–4 °C
Density:1.26 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2206
ADR:6.1,II
:GHS06
:GHS08
Frequently Bought Together