N,N-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1231967-65-9
:
Formula:C₁₄H₂₃BClNO₂
MDL Number:MFCD11052310
:283.61 g/mol
Storage Temperature:Ambient
Frequently Bought Together