Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boc-DL-beta-Aib-OH
Boc-DL-β-Aib-OH
Boc-DL-β-Aib-OH
APOSOR309427-250MG
:
Apollo Scientific
:
Boc-DL-β-Aib-OH
APOSOR309427-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
203.24 g/mol
:
88…90 °C
:
GHS07
:
Ambient
:
16948-10-0
:
C₉H₁₇NO₄
:
MFCD02094137