Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1-Benzylazetidine-2,4-diyl)dimethanol
(1-Benzylazetidine-2,4-diyl)dimethanol
(1-Benzylazetidine-2,4-diyl)dimethanol
APOSOR309011-1G
:
Apollo Scientific
:
(1-Benzylazetidine-2,4-diyl)dimethanol
APOSOR309011-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
207.27 g/mol
:
GHS07
:
Refrigerator
:
127310-66-1
:
1263280-69-8
:
C₁₂H₁₇NO₂
:
MFCD17215364