Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Chloro-6-methoxyquinoline

2-Chloro-6-methoxyquinoline

Irritant
Supplier:   Apollo Scientific
2-Chloro-6-methoxyquinoline
Supplier:  Apollo Scientific
Certificates
2-Chloro-6-methoxyquinoline

5 Options Available Below

Product Details & Documents
  • CAS Number:13676-02-3
  • Formula:C₁₀H₈ClNO
  • MDL Number:MFCD00799212
  • Molecular Weight:193.63
  • Melting Pt:108…109 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - £16.50
    £16.50
    1 * 250 mg
       £16.50

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - £40.70
    £40.70
    1 * 1 g
       £40.70

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £150.00
    £150.00
    1 * 5 g
       £150.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £676.00
    £676.00
    1 * 25 g
       £676.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £2,650.00
    £2,650.00
    1 * 100 g
       £2,650.00

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.