
2,3-Dimethylbenzylamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51586-20-0
:
Formula:C₉H₁₃N
MDL Number:MFCD00052680
:135.21 g/mol
Boiling Pt:118…120 °C/15mmHg
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, dehydrated (max. 0.01% H₂O) ≥99%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
1,2,3,4-Tetrahydronaphthalene for synthesis 1 * 25 kg
Supplier: Merck
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
n-Hexane 99+% for analysis
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane ≥99.0%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9%, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific