
(4-tert-Butylphenyl)methanesulphonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:519056-61-2
:
Formula:C₁₁H₁₅ClO₂S
MDL Number:MFCD04115413
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together