3,5-Dimethyl-p-anisic acid 98%
Catalog # APOSOR29323-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:180.2 g/mol
Storage Temperature:Ambient
CAS Number:21553-46-8
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00020309