Order Entry
United Kingdom
ContactUsLinkComponent
Propagyl amine hydrochloride
Propagyl amine hydrochloride
Catalog # APOSOR27447-25G
Supplier:  Apollo Scientific
CAS Number:  
Propagyl amine hydrochloride
Catalog # APOSOR27447-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :91.54 g/mol
Storage Temperature:Ambient
CAS Number:15430-52-1
Formula:C₃H₆ClN
MDL Number:MFCD00012907