Order Entry
United Kingdom
ContactUsLinkComponent
Ethyl-1-cyclopropyl-4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
Ethyl-1-cyclopropyl-4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
Catalog # APOSOR25975-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-1-cyclopropyl-4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
Catalog # APOSOR25975-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :235.29 g/mol
Storage Temperature:Ambient
CAS Number:423768-53-0
Formula:C₁₃H₁₇NO₃
MDL Number:MFCD03659714