
1-Boc-5-aminoindole 95%
Catalog # APOSOR2566-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.28 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:166104-20-7
Formula:C₁₃H₁₆N₂O₂
MDL Number:MFCD04114746