
2,2-Dimethyl-2H-chromene-6-carbonitrile 97%
Catalog # APOSOR25515-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:185.23 g/mol
Melting Pt:45…49 °C
:GHS07
Storage Temperature:Ambient
CAS Number:33143-29-2
Formula:C₁₂H₁₁NO
MDL Number:MFCD00084978