Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Phenyl-1H-pyrazole-5-carbaldehyde
1-Phenyl-1H-pyrazole-5-carbaldehyde
1-Phenyl-1H-pyrazole-5-carbaldehyde
APOSOR23388-250MG
:
Apollo Scientific
:
1-Phenyl-1H-pyrazole-5-carbaldehyde
APOSOR23388-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
172.1800 g/mol
:
GHS07
:
Ambient
:
132274-70-5
:
C₁₀H₈N₂O
:
MFCD00103497