
1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid
Catalog # APOSOR23236-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:154.17 g/mol
Melting Pt:215…219 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1125-29-7
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD00159648