
Quinoxaline-6-carbonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:258503-93-4
:
Formula:C₉H₅ClN₂O
MDL Number:MFCD02682002
:192.61 g/mol
Melting Pt:119 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together