Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Bromothiazole

2-Bromothiazole

Irritant
Supplier:   Apollo Scientific
2-Bromothiazole
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:3034-53-5
  • Formula:C₃H₂BrNS
  • MDL Number:MFCD00005316
  • Molecular Weight:164.03 g/mol
  • Boiling Pt:171 °C
  • Density:1.82 g/cm³ (25 °C)
  • Flash Pt:67 °C
  • Storage Temperature:Refrigerator
  • EINECS:221-229-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £16.50
    £16.50
    1 * 25 g
       £16.50

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £18.70
    £18.70
    1 * 100 g
       £18.70

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £81.40
    £81.40
    1 * 500 g
       £81.40

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.