Benzothiazol-2-ylmethanol
Catalog # APOSOR23091-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:165.21 g/mol
Melting Pt:95…100 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:37859-42-0
Formula:C₈H₇NOS
MDL Number:MFCD00226293