
Isonicotinic acid 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:55-22-1
:
Formula:C₆H₅NO₂
MDL Number:MFCD00006429
:123.11 g/mol
Boiling Pt:260 °C/15mmHg
Melting Pt:310…315 °C (subl.)
Storage Temperature:Ambient
EINECS:200-228-2
:GHS07
Frequently Bought Together