
2'-Hydroxy-4'-methylacetophenone 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6921-64-8
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00100559
:150.18 g/mol
Boiling Pt:245 °C
Density:1.1 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together