Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

2-Bromo-4-chlorophenol 97%

Irritant
New Product
Supplier:   Apollo Scientific
2-Bromo-4-chlorophenol 97%
Supplier:  Apollo Scientific
New Product
Certificates
2-Bromo-4-chlorophenol 97%

2 Options Available Below

Product Details & Documents
  • CAS Number:695-96-5
  • Formula:C₆H₄BrClO
  • MDL Number:MFCD00002319
  • Molecular Weight:207.45 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £42.90
    £42.90
    1 * 100 g
       £42.90

    Specifications:
    New Product
    New Product
  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £192.00
    £192.00
    1 * 500 g
       £192.00