Order Entry
United Kingdom
ContactUsLinkComponent
3-(4-Nitrophenoxy)benzaldehyde
3-(4-Nitrophenoxy)benzaldehyde
Catalog # APOSOR200044-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Nitrophenoxy)benzaldehyde
Catalog # APOSOR200044-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.22 g/mol
Melting Pt:84 °C
Storage Temperature:Ambient
CAS Number:17076-72-1
Formula:C₁₃H₉NO₄
MDL Number:MFCD18207479