
1-Benzhydryl-3-azetidinol hydrochloride 95%
Catalog # APOSOR1872-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:275.78 g/mol
:GHS05
Storage Temperature:Ambient
CAS Number:90604-02-7
Formula:C₁₆H₁₇NO·HCl
MDL Number:MFCD00209447