Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Cyclohexyloxy)isonicotinic acid
2-(Cyclohexyloxy)isonicotinic acid
2-(Cyclohexyloxy)isonicotinic acid
APOSOR18621-1G
:
Apollo Scientific
:
2-(Cyclohexyloxy)isonicotinic acid
APOSOR18621-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
221.26 g/mol
:
Ambient
:
1019353-19-5
:
C₁₂H₁₅NO₃
:
MFCD09258836