4-Bromoindole-3-carboxyaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:98600-34-1
:
Formula:C₉H₆BrNO
MDL Number:MFCD05864695
Melting Pt:179 °C
:224.06 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together