
Ethyl-1,3-benzothiazole-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:32137-76-1
:
Formula:C₁₀H₉NO₂S
MDL Number:MFCD00848360
:207.25 g/mol
Melting Pt:68…72 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together