Order Entry
United Kingdom
ContactUsLinkComponent
5-Bromopyridine-2-carbonyl chloride, Technical Grade
Corrosive
5-Bromopyridine-2-carbonyl chloride, Technical Grade
Catalog # APOSOR15012-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
5-Bromopyridine-2-carbonyl chloride, Technical Grade
Catalog # APOSOR15012-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.45 g/mol
Melting Pt:120 °C (dec.)
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
CAS Number:137178-88-2
Formula:C₆H₃BrClNO
MDL Number:MFCD07368290