

Oxazole 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:288-42-6
:
Formula:C₃H₃NO
MDL Number:MFCD00009751
:69.06 g/mol
Boiling Pt:69…70 °C
Melting Pt:–87 °C
Density:1.05 g/cm³ (25 °C)
Flash Pt:19 °C
Storage Temperature:Refrigerator
EINECS:206-020-8
UN:1993
ADR:3,II
:GHS02
:GHS05
Frequently Bought Together