
3-(4-Methylphenyl)-1H-pyrazole-4-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:350988-62-4
:
Formula:C₁₁H₁₀N₂O
MDL Number:MFCD01133694
Melting Pt:123…125 °C
:186.22 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together