Order Entry
United Kingdom
ContactUsLinkComponent
7-Amino-2H-1,4-benzoxazin-3(4H)-one
7-Amino-2H-1,4-benzoxazin-3(4H)-one
Catalog # APOSOR13747-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Amino-2H-1,4-benzoxazin-3(4H)-one
Catalog # APOSOR13747-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.17 g/mol
Melting Pt:220…222 °C
Storage Temperature:Ambient
CAS Number:26215-14-5
Formula:C₈H₈N₂O₂
MDL Number:MFCD03425794