
Methyl isonicotinate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2459-09-8
:
Formula:C₇H₇NO₂
MDL Number:MFCD00006427
:137.14 g/mol
Boiling Pt:207…209 °C
Melting Pt:8…8.5 °C
Density:1.161 g/cm³ (25 °C)
Flash Pt:82 °C
EINECS:219-546-8
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together