Order Entry
United Kingdom
ContactUsLinkComponent
4-Hydroxy-2,6-dimethylbenzaldehyde 98%
Irritant
4-Hydroxy-2,6-dimethylbenzaldehyde 98%
Catalog # APOSOR12903-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Hydroxy-2,6-dimethylbenzaldehyde 98%
Catalog # APOSOR12903-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :150.1700 g/mol
Melting Pt:190…196 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:70547-87-4
Formula:C₉H₁₀O₂
MDL Number:MFCD00016979